Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAGGTCATTCATTATGAAGTGTAC[C/T]GCGCGGGAGTGGCTCAGAGTAACCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
CRISPLD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CRISPLD1 - cysteine rich secretory protein LCCL domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286777.1 | 522 | Intron | NP_001273706.1 | |||
NM_001286778.1 | 522 | Intron | NP_001273707.1 | |||
NM_031461.5 | 522 | Silent Mutation | ACC,ACT | T,T 4 | NP_113649.1 |