Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGCCTCTGGTCAATGTAGTTAAC[C/T]ACACTTGTCTAGATGTGGAGGAGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
TMEM206 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TMEM206 - transmembrane protein 206 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001198862.1 | 1034 | Silent Mutation | GTA,GTG | V,V 325 | NP_001185791.1 | |
NM_018252.2 | 1034 | Silent Mutation | GTA,GTG | V,V 264 | NP_060722.2 | |
XM_005273177.3 | 1034 | Silent Mutation | GTA,GTG | V,V 187 | XP_005273234.1 | |
XM_011509717.2 | 1034 | Silent Mutation | GTA,GTG | V,V 349 | XP_011508019.1 | |
XM_011509718.2 | 1034 | Nonsense Mutation | TAG,TGG | *,W 301 | XP_011508020.1 | |
XM_017001634.1 | 1034 | Silent Mutation | GTA,GTG | V,V 279 | XP_016857123.1 | |
XM_017001635.1 | 1034 | Silent Mutation | GTA,GTG | V,V 194 | XP_016857124.1 | |
XM_017001636.1 | 1034 | Silent Mutation | GTA,GTG | V,V 187 | XP_016857125.1 | |
XM_017001637.1 | 1034 | Nonsense Mutation | TAG,TGG | *,W 216 | XP_016857126.1 |