Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTAAATCATTATAACAGCTCCTGCT[A/G]ATGTCTGACGAATCTCTCATATTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300390 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CHM PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CHM - CHM, Rab escort protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000390.3 | 1709 | Silent Mutation | ATC,ATT | I,I 560 | NP_000381.1 | |
NM_001145414.3 | 1709 | Intron | NP_001138886.1 | |||
NM_001320959.1 | 1709 | Silent Mutation | ATC,ATT | I,I 412 | NP_001307888.1 | |
XM_017029242.1 | 1709 | Silent Mutation | ATC,ATT | I,I 560 | XP_016884731.1 | |
XM_017029243.1 | 1709 | Silent Mutation | ATC,ATT | I,I 412 | XP_016884732.1 | |
XM_017029244.1 | 1709 | Silent Mutation | ATC,ATT | I,I 412 | XP_016884733.1 | |
XM_017029245.1 | 1709 | Silent Mutation | ATC,ATT | I,I 412 | XP_016884734.1 | |
XM_017029246.1 | 1709 | Silent Mutation | ATC,ATT | I,I 412 | XP_016884735.1 |