Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATGTAATGAAACACTAACTACAAG[C/T]TAGTTCTATATCACTAAAACTCAGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616142 MIM: 603962 | ||||||||||||||||||||
Literature Links: |
FAM98B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FAM98B - family with sequence similarity 98 member B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RASGRP1 - RAS guanyl releasing protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001128602.1 | 3996 | UTR 3 | NP_001122074.1 | |||
NM_001306086.1 | 3996 | UTR 3 | NP_001293015.1 | |||
NM_005739.3 | 3996 | UTR 3 | NP_005730.2 | |||
XM_005254114.2 | 3996 | UTR 3 | XP_005254171.1 | |||
XM_011521151.2 | 3996 | UTR 3 | XP_011519453.1 | |||
XM_017021860.1 | 3996 | UTR 3 | XP_016877349.1 |