Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTCAGAGCAAAACAGGAAGAAGGC[A/G]GATCTGAAAGGAAGCAACAGAGTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605005 MIM: 163906 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GALNT7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GALNT7 - polypeptide N-acetylgalactosaminyltransferase 7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HMGB2 - high mobility group box 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130688.1 | 433 | Silent Mutation | TCC,TCT | S,S 100 | NP_001124160.1 | |
NM_001130689.1 | 433 | Silent Mutation | TCC,TCT | S,S 100 | NP_001124161.1 | |
NM_002129.3 | 433 | Silent Mutation | TCC,TCT | S,S 100 | NP_002120.1 |
LOC105377538 - uncharacterized LOC105377538 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |