Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATAAAATTGAAGATGTTCTTCAAA[C/T]GAATATCGAAGAATGTTTGGGTGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610200 MIM: 605927 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MRPL13 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MRPL13 - mitochondrial ribosomal protein L13 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MTBP - MDM2 binding protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022045.4 | 414 | Missense Mutation | ACG,ATG | T,M 115 | NP_071328.2 | |
XM_011516962.2 | 414 | Missense Mutation | ACG,ATG | T,M 115 | XP_011515264.1 | |
XM_011516963.2 | 414 | Missense Mutation | ACG,ATG | T,M 115 | XP_011515265.1 |