Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCTGGGGACCATGCAGAGCTACAT[A/G]GATGCCTCCCTTATCTCCCTCATTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
PPRC1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PPRC1 - peroxisome proliferator-activated receptor gamma, coactivator-related 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001288727.1 | 327 | Missense Mutation | ATA,ATG | I,M 98 | NP_001275656.1 | |
NM_001288728.1 | 327 | Nonsense Mutation | TAG,TGG | *,W 12 | NP_001275657.1 | |
NM_015062.4 | 327 | Missense Mutation | ATA,ATG | I,M 98 | NP_055877.3 | |
XM_005269656.3 | 327 | Missense Mutation | ATA,ATG | I,M 98 | XP_005269713.1 | |
XM_006717731.3 | 327 | Missense Mutation | ATA,ATG | I,M 98 | XP_006717794.1 | |
XM_011539549.2 | 327 | Missense Mutation | ATA,ATG | I,M 98 | XP_011537851.1 | |
XM_011539551.2 | 327 | Missense Mutation | ATA,ATG | I,M 98 | XP_011537853.1 | |
XM_011539552.2 | 327 | Missense Mutation | ATA,ATG | I,M 98 | XP_011537854.1 | |
XM_017015982.1 | 327 | Missense Mutation | ATA,ATG | I,M 98 | XP_016871471.1 | |
XM_017015983.1 | 327 | Missense Mutation | ATA,ATG | I,M 98 | XP_016871472.1 | |
XM_017015984.1 | 327 | Missense Mutation | ATA,ATG | I,M 98 | XP_016871473.1 | |
XM_017015985.1 | 327 | Missense Mutation | ATA,ATG | I,M 98 | XP_016871474.1 |