Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAGTTTTGTTCGATTCCGTCCCAC[A/G]TTCAGTTTGGGTACTGCAGTCTCGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300410 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AMOT PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AMOT - angiomotin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001113490.1 | 2375 | Missense Mutation | CGT,TGT | R,C 868 | NP_001106962.1 | |
NM_133265.2 | 2375 | Missense Mutation | CGT,TGT | R,C 459 | NP_573572.1 | |
XM_005262087.1 | 2375 | Missense Mutation | CGT,TGT | R,C 868 | XP_005262144.1 | |
XM_005262090.1 | 2375 | Missense Mutation | CGT,TGT | R,C 459 | XP_005262147.1 | |
XM_011530875.2 | 2375 | Missense Mutation | CGT,TGT | R,C 868 | XP_011529177.1 | |
XM_017029289.1 | 2375 | Missense Mutation | CGT,TGT | R,C 459 | XP_016884778.1 |
MIR4329 - microRNA 4329 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |