Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGTTCCTGACGGACCGGCGCATCC[A/G]CGAGGACTACCCGCAGGTGCAGATC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
RIMKLA PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RIMKLA - ribosomal modification protein rimK like family member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_173642.3 | 179 | Missense Mutation | CAC,CGC | H,R 14 | NP_775913.2 | |
XM_006710585.3 | 179 | Missense Mutation | CAC,CGC | H,R 14 | XP_006710648.1 | |
XM_017001082.1 | 179 | UTR 5 | XP_016856571.1 |