Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTACTGGCCCGTCCGACGCAAGGTC[A/C]ACCGCGTCTTCAGCACCTGAGGATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603445 MIM: 608746 MIM: 610822 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KHSRP PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KHSRP - KH-type splicing regulatory protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC25A23 - solute carrier family 25 member 23 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC25A41 - solute carrier family 25 member 41 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321298.1 | 521 | Missense Mutation | TGG,TTG | W,L 76 | NP_001308227.1 | |
NM_173637.3 | 521 | Missense Mutation | TGG,TTG | W,L 214 | NP_775908.2 | |
XM_011527926.1 | 521 | Missense Mutation | TGG,TTG | W,L 150 | XP_011526228.1 |