Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGTCATTTACCTTCATTGACTTC[A/G]TCATTTTCTCTGTCCACCTGGGCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
TRPC7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TRPC7 - transient receptor potential cation channel subfamily C member 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001167576.1 | 2280 | Silent Mutation | GAC,GAT | D,D 686 | NP_001161048.1 | |
NM_001167577.1 | 2280 | Silent Mutation | GAC,GAT | D,D 741 | NP_001161049.1 | |
NM_020389.2 | 2280 | Silent Mutation | GAC,GAT | D,D 802 | NP_065122.1 |