Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCCCCAATGGGCTGAAGCTCATCT[C/T]GGAAGTGATTGGGGAGCGCCTCGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614478 MIM: 138420 MIM: 601735 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
COX14 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
COX14 - COX14, cytochrome c oxidase assembly factor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GPD1 - glycerol-3-phosphate dehydrogenase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001257199.1 | 561 | Missense Mutation | TCG,TTG | S,L 110 | NP_001244128.1 | |
NM_005276.3 | 561 | Missense Mutation | TCG,TTG | S,L 133 | NP_005267.2 |
SMARCD1 - SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily d, member 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |