Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCGGCTTGCCTACTTCTGTACACG[A/G]AGACTGGCCCTGCAGCTGGATGGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 308380 MIM: 300188 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IL2RG PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IL2RG - interleukin 2 receptor subunit gamma | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MED12 - mediator complex subunit 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005120.2 | 1108 | Silent Mutation | CGA,CGG | R,R 303 | NP_005111.2 |