Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGATGTGAGCACCGTTTAAAAGCT[G/T]TGCCACATTCTTCACATTTATAGCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF682 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF682 - zinc finger protein 682 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001077349.1 | 1606 | Intron | NP_001070817.1 | |||
NM_033196.2 | 1606 | Intron | NP_149973.1 | |||
XM_017027455.1 | 1606 | Missense Mutation | AAA,ACA | K,T 466 | XP_016882944.1 | |
XM_017027456.1 | 1606 | Intron | XP_016882945.1 |