Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGAGATGGCGGAACAGCACGGAGC[A/G]CCCGAACAGGCTGCAGCTGGCAAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 123630 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CRYBB3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CRYBB3 - crystallin beta B3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004076.4 | 639 | Silent Mutation | GCA,GCG | A,A 7 | NP_004067.1 | |
XM_011529902.2 | 639 | Silent Mutation | GCA,GCG | A,A 63 | XP_011528204.1 | |
XM_017028599.1 | 639 | Silent Mutation | GCA,GCG | A,A 63 | XP_016884088.1 |
KIAA1671 - KIAA1671 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |