Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCCACCATGCTGCAGTGTAGACCC[A/G]CACAGGAGTTCAGCTTCGGGCCCCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615168 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AMZ1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AMZ1 - archaelysin family metallopeptidase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001284355.2 | 332 | Missense Mutation | ACA,GCA | T,A 7 | NP_001271284.1 | |
NM_001321766.1 | 332 | Missense Mutation | ACA,GCA | T,A 7 | NP_001308695.1 | |
NM_133463.3 | 332 | Missense Mutation | ACA,GCA | T,A 7 | NP_597720.1 | |
XM_011515150.2 | 332 | Missense Mutation | ACA,GCA | T,A 7 | XP_011513452.1 | |
XM_011515151.2 | 332 | Missense Mutation | ACA,GCA | T,A 7 | XP_011513453.1 | |
XM_017011774.1 | 332 | Missense Mutation | ACA,GCA | T,A 7 | XP_016867263.1 | |
XM_017011775.1 | 332 | Missense Mutation | ACA,GCA | T,A 7 | XP_016867264.1 | |
XM_017011776.1 | 332 | Missense Mutation | ACA,GCA | T,A 7 | XP_016867265.1 |