Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAATGTTTGGAAGCCAAAACCAGA[A/G]CAGACCAGATTTTTTACTTTCTGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608570 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AHNAK2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AHNAK2 - AHNAK nucleoprotein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_138420.2 | 17053 | Missense Mutation | CTC,TTC | L,F 5645 | NP_612429.2 | |
XM_005267299.1 | 17053 | Missense Mutation | CTC,TTC | L,F 5545 | XP_005267356.1 |
PLD4 - phospholipase D family member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308174.1 | 17053 | Intron | NP_001295103.1 | |||
NM_138790.3 | 17053 | Intron | NP_620145.2 | |||
XM_011536411.2 | 17053 | Intron | XP_011534713.1 | |||
XM_017020965.1 | 17053 | Intron | XP_016876454.1 |