Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGCAGCTTCACGGACTTCTCCACG[C/T]TGCCCTCGGGCGCCTTCAAAGCCGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609719 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LHFPL3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LHFPL3 - lipoma HMGIC fusion partner-like 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_199000.2 | 299 | Silent Mutation | CTG,TTG | L,L 100 | NP_945351.1 | |
XM_005250327.1 | 299 | Silent Mutation | CTG,TTG | L,L 100 | XP_005250384.1 |