Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATGTCATCTACTGCTACCACAAAC[A/G]GTCGAGAACCACTGTTGGGGGAAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300862 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACOT9 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ACOT9 - acyl-CoA thioesterase 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001033583.2 | 1177 | Missense Mutation | CCG,CTG | P,L 335 | NP_001028755.2 | |
NM_001037171.1 | 1177 | Missense Mutation | CCG,CTG | P,L 344 | NP_001032248.1 | |
XM_005274472.2 | 1177 | Missense Mutation | CCG,CTG | P,L 275 | XP_005274529.1 |