Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCCACCAGGTAGCCCTCCGGGCTA[A/G]GTCCCCTTCCTCCTAGTGTGCTTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601336 MIM: 611857 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC142 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCDC142 - coiled-coil domain containing 142 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032779.3 | 2507 | Missense Mutation | CCT,CTT | P,L 705 | NP_116168.3 |
MOGS - mannosyl-oligosaccharide glucosidase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MRPL53 - mitochondrial ribosomal protein L53 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TTC31 - tetratricopeptide repeat domain 31 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |