Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTGAGCGAGGGCGGCCTGGGGACC[A/G]CCGGGTGGCTGTGTCGTCATTAGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601517 MIM: 605093 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATXN2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ATXN2 - ataxin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001310121.1 | 3268 | Silent Mutation | GGC,GGT | G,G 1035 | NP_001297050.1 | |
NM_001310123.1 | 3268 | Silent Mutation | GGC,GGT | G,G 968 | NP_001297052.1 | |
NM_002973.3 | 3268 | Silent Mutation | GGC,GGT | G,G 1275 | NP_002964.3 |
SH2B3 - SH2B adaptor protein 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |