Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTACTTGAAGGCAGGTGCTTCTCAC[C/T]GCTGCTCTGCTTTACAGGTACATTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
KIAA1210 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KIAA1210 - KIAA1210 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020721.1 | 3670 | Missense Mutation | AGT,GGT | S,G 1361 | NP_065772.1 | |
XM_006724774.3 | 3670 | Missense Mutation | AGT,GGT | S,G 1185 | XP_006724837.1 | |
XM_017029688.1 | 3670 | Missense Mutation | AGT,GGT | S,G 1200 | XP_016885177.1 | |
XM_017029689.1 | 3670 | Missense Mutation | AGT,GGT | S,G 1134 | XP_016885178.1 |