Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCCATGTTCTCTTTCCCGAGGCTG[C/T]CCTGGGGCGGAGGTGACCTTGTGCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613368 MIM: 610831 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CARNS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CARNS1 - carnosine synthase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001166222.1 | 362 | Silent Mutation | TGC,TGT | C,C 93 | NP_001159694.1 | |
NM_020811.1 | 362 | UTR 5 | NP_065862.1 | |||
XM_011545189.2 | 362 | Silent Mutation | TGC,TGT | C,C 93 | XP_011543491.1 | |
XM_011545191.2 | 362 | Silent Mutation | TGC,TGT | C,C 109 | XP_011543493.1 | |
XM_017018054.1 | 362 | Silent Mutation | TGC,TGT | C,C 109 | XP_016873543.1 | |
XM_017018055.1 | 362 | Intron | XP_016873544.1 |
TBC1D10C - TBC1 domain family member 10C | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |