Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAACCAGAGTAAGATATACCATGTA[C/G]AGAAGTAGAAGGCCCAAAATGGAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
TMEM9B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TMEM9B - TMEM9 domain family member B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286094.1 | 811 | Silent Mutation | CTC,CTG | L,L 43 | NP_001273023.1 | |
NM_001286095.1 | 811 | Silent Mutation | CTC,CTG | L,L 43 | NP_001273024.1 | |
NM_020644.2 | 811 | Silent Mutation | CTC,CTG | L,L 117 | NP_065695.1 |