Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGGGACCCAACTGGGAGACAAATT[C/G]GACTCGTTCATTAAACCACCTGAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601118 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CAMLG PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CAMLG - calcium modulating ligand | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001745.3 | 1036 | Missense Mutation | TTC,TTG | F,L 114 | NP_001736.1 | |
XM_011543653.1 | 1036 | Missense Mutation | TTC,TTG | F,L 114 | XP_011541955.1 |