Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTGGCCTACCTTGCCCTTGCGGAA[A/G]AGAGCCTTGATGTTGTCTGGCTGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604840 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FKBP8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FKBP8 - FK506 binding protein 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308373.1 | 1124 | Silent Mutation | CTC,CTT | L,L 309 | NP_001295302.1 | |
NM_012181.4 | 1124 | Silent Mutation | CTC,CTT | L,L 310 | NP_036313.3 | |
XM_011527863.1 | 1124 | Silent Mutation | CTC,CTT | L,L 310 | XP_011526165.1 | |
XM_011527864.1 | 1124 | Silent Mutation | CTC,CTT | L,L 310 | XP_011526166.1 |