Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCGCTGACGTCGCCGCCCAGATGG[C/T]CTCCAGGCTGACCCTGCTGACCCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606860 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SERPING1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SERPING1 - serpin family G member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000062.2 | 44 | Missense Mutation | GCC,GTC | A,V 2 | NP_000053.2 | |
NM_001032295.1 | 44 | Missense Mutation | GCC,GTC | A,V 2 | NP_001027466.1 |