Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAAAGGAGGATGGCAGCAAACAAG[G/T]GTCTGTGCACAGGAGTCAGAGAGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609093 MIM: 615180 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FBXO15 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FBXO15 - F-box protein 15 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TIMM21 - translocase of inner mitochondrial membrane 21 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014177.2 | 530 | Missense Mutation | GGG,GTG | G,V 83 | NP_054896.2 | |
XM_005266689.3 | 530 | Missense Mutation | AGG,AGT | R,S 28 | XP_005266746.1 |