Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAATAAAACTGTGAACAGCTGGACA[A/G]GAACACTGGTCTACTCGCCGAACTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
SBSPON PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SBSPON - somatomedin B and thrombospondin type 1 domain containing | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_153225.3 | 967 | Silent Mutation | TCC,TCT | S,S 254 | NP_694957.3 | |
XM_017013145.1 | 967 | Silent Mutation | TCC,TCT | S,S 192 | XP_016868634.1 |