Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCGAGGGGGCCGGCGTGCTGGGCCT[C/G]AGTGCGTCCGCCGAGTGCCACCTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600010 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CFAP61 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CFAP61 - cilia and flagella associated protein 61 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
INSM1 - INSM transcriptional repressor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002196.2 | 1449 | Silent Mutation | CTC,CTG | L,L 434 | NP_002187.1 |