Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGATTCCTGGACAAAAGGAAATCTG[A/C]CAGCTGACCTCCTAAAATTGTACAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611049 | ||||||||||||||||||||
Literature Links: |
SLC17A2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SLC17A2 - solute carrier family 17 member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286123.1 | 1530 | Missense Mutation | GCA,TCA | A,S 333 | NP_001273052.1 | |
NM_001286125.1 | 1530 | Missense Mutation | GCA,TCA | A,S 333 | NP_001273054.1 | |
NM_005835.3 | 1530 | Missense Mutation | GCA,TCA | A,S 333 | NP_005826.1 | |
XM_005248784.2 | 1530 | Missense Mutation | GCA,TCA | A,S 333 | XP_005248841.1 | |
XM_006714949.3 | 1530 | Missense Mutation | GCA,TCA | A,S 333 | XP_006715012.1 | |
XM_006714950.2 | 1530 | Missense Mutation | GCA,TCA | A,S 310 | XP_006715013.1 | |
XM_006714951.1 | 1530 | Missense Mutation | GCA,TCA | A,S 333 | XP_006715014.1 | |
XM_017010159.1 | 1530 | Missense Mutation | GCA,TCA | A,S 310 | XP_016865648.1 | |
XM_017010160.1 | 1530 | Missense Mutation | GCA,TCA | A,S 333 | XP_016865649.1 |