Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCCAGAGCAAGTTGTTTGAATTCA[C/T]AGTGCTTCTCCTCTAGAATGTCTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609369 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATL3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ATL3 - atlastin GTPase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001290048.1 | 1456 | Missense Mutation | TAT,TGT | Y,C 370 | NP_001276977.1 | |
NM_015459.4 | 1456 | Missense Mutation | TAT,TGT | Y,C 388 | NP_056274.3 | |
XM_006718493.1 | 1456 | Missense Mutation | TAT,TGT | Y,C 369 | XP_006718556.1 |