Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTCTGGCCCATTCTTCTCTCCTCC[A/G]GTTTCTGGCCCAACTGGAGTCCCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610393 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GON4L PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GON4L - gon-4 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282856.1 | 4489 | Silent Mutation | ACC,ACT | T,T 1450 | NP_001269785.1 | |
NM_001282858.1 | 4489 | Silent Mutation | ACC,ACT | T,T 1450 | NP_001269787.1 | |
NM_001282860.1 | 4489 | Silent Mutation | ACC,ACT | T,T 1450 | NP_001269789.1 | |
NM_001282861.1 | 4489 | Silent Mutation | ACC,ACT | T,T 1450 | NP_001269790.1 | |
NM_032292.5 | 4489 | Silent Mutation | ACC,ACT | T,T 1450 | NP_115668.4 | |
XM_005245284.3 | 4489 | Silent Mutation | ACC,ACT | T,T 1278 | XP_005245341.1 | |
XM_005245286.3 | 4489 | Silent Mutation | ACC,ACT | T,T 646 | XP_005245343.1 | |
XM_006711393.3 | 4489 | Silent Mutation | ACC,ACT | T,T 1450 | XP_006711456.1 | |
XM_006711394.3 | 4489 | Silent Mutation | ACC,ACT | T,T 1450 | XP_006711457.1 | |
XM_011509658.2 | 4489 | Silent Mutation | ACC,ACT | T,T 1421 | XP_011507960.1 | |
XM_011509659.2 | 4489 | Silent Mutation | ACC,ACT | T,T 1278 | XP_011507961.1 |