Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTACAACCCTTATGCTGCCCTTTCC[A/G]TAGAGCAGCAGAGGCAGAAGCTGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
DHX35 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DHX35 - DEAH-box helicase 35 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001190809.1 | 169 | Missense Mutation | ATA,GTA | I,V 47 | NP_001177738.1 | |
NM_021931.3 | 169 | Missense Mutation | ATA,GTA | I,V 47 | NP_068750.2 | |
XM_006723848.3 | 169 | Intron | XP_006723911.1 | |||
XM_011528972.1 | 169 | Missense Mutation | ATA,GTA | I,V 47 | XP_011527274.1 | |
XM_011528974.1 | 169 | Missense Mutation | ATA,GTA | I,V 47 | XP_011527276.1 | |
XM_017028004.1 | 169 | Intron | XP_016883493.1 |