Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATATCCATAACAAATTTTTCTTCA[A/C]AATGTCTTTTATATATTTCTTGAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
LRRIQ3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LRRIQ3 - leucine rich repeats and IQ motif containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001105659.1 | 1141 | Missense Mutation | TGT,TTT | C,F 582 | NP_001099129.1 | |
NM_001322315.1 | 1141 | Missense Mutation | GTG,TTG | V,L 423 | NP_001309244.1 | |
XM_011540658.1 | 1141 | Missense Mutation | TGT,TTT | C,F 474 | XP_011538960.1 | |
XM_011540659.1 | 1141 | Missense Mutation | TGT,TTT | C,F 350 | XP_011538961.1 | |
XM_011540660.1 | 1141 | Missense Mutation | TGT,TTT | C,F 294 | XP_011538962.1 | |
XM_011540662.2 | 1141 | Missense Mutation | TGT,TTT | C,F 294 | XP_011538964.1 | |
XM_011540663.2 | 1141 | Intron | XP_011538965.1 | |||
XM_017000286.1 | 1141 | Intron | XP_016855775.1 | |||
XM_017000287.1 | 1141 | Intron | XP_016855776.1 | |||
XM_017000288.1 | 1141 | Intron | XP_016855777.1 |