Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGAAGCTCATCGAGTCAAGACTGC[C/G]AAACTACAGCTGGTGGAACAGCAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600888 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARHGEF5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARHGEF5 - Rho guanine nucleotide exchange factor 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005435.3 | 4882 | Silent Mutation | GCC,GCG | A,A 1588 | NP_005426.2 | |
XM_017012623.1 | 4882 | Intron | XP_016868112.1 |