Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAAGAAGCACCTATTGAATCACTG[C/G]CTGCAAAGAAGGAAGCAGCAGCCAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605011 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADAMTS3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ADAMTS3 - ADAM metallopeptidase with thrombospondin type 1 motif 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014243.2 | 4083 | Missense Mutation | GCC,GGC | A,G 1172 | NP_055058.2 | |
XM_011532421.2 | 4083 | Missense Mutation | GCC,GGC | A,G 1153 | XP_011530723.1 | |
XM_011532422.2 | 4083 | Missense Mutation | GCC,GGC | A,G 1144 | XP_011530724.1 |