Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTTTGCCACACAGCTCATCCGGAA[C/G]GGGGAGACCATCTTCGTAGAACGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
NOTO PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NOTO - notochord homeobox | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PRADC1 - protease associated domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SMYD5 - SMYD family member 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006062.2 | 1881 | Missense Mutation | AAC,AAG | N,K 44 | NP_006053.2 | |
XM_006711918.3 | 1881 | UTR 5 | XP_006711981.1 | |||
XM_017003163.1 | 1881 | Missense Mutation | AAC,AAG | N,K 56 | XP_016858652.1 |