Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAAATCTATCCATGACACCTATTA[C/T]GAAAGTTACATGTGAAGAATTCAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613813 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MDM1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MDM1 - Mdm1 nuclear protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001205028.1 | 2421 | UTR 3 | NP_001191957.1 | |||
NM_001205029.1 | 2421 | Intron | NP_001191958.1 | |||
NM_017440.4 | 2421 | UTR 3 | NP_059136.2 | |||
NM_020128.2 | 2421 | Intron | NP_064513.1 | |||
XM_005269026.1 | 2421 | UTR 3 | XP_005269083.1 | |||
XM_011538569.1 | 2421 | Intron | XP_011536871.1 | |||
XM_017019646.1 | 2421 | UTR 3 | XP_016875135.1 | |||
XM_017019647.1 | 2421 | UTR 3 | XP_016875136.1 | |||
XM_017019648.1 | 2421 | UTR 3 | XP_016875137.1 | |||
XM_017019649.1 | 2421 | UTR 3 | XP_016875138.1 | |||
XM_017019650.1 | 2421 | UTR 3 | XP_016875139.1 |