Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGATGATGGCCACTCCGAACCAGAC[A/C]GCCTGTAATGCAGAGTCACCAGTGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615695 | ||||||||||||||||||||
Literature Links: |
HEXIM2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HEXIM2 - hexamethylene bisacetamide inducible 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303436.1 | 1143 | Silent Mutation | ACA,ACC | T,T 30 | NP_001290365.1 | |
NM_001303437.1 | 1143 | Silent Mutation | ACA,ACC | T,T 8 | NP_001290366.1 | |
NM_001303438.1 | 1143 | Silent Mutation | ACA,ACC | T,T 8 | NP_001290367.1 | |
NM_001303439.1 | 1143 | Silent Mutation | ACA,ACC | T,T 8 | NP_001290368.1 | |
NM_001303440.1 | 1143 | Silent Mutation | ACA,ACC | T,T 8 | NP_001290369.1 | |
NM_001303441.1 | 1143 | Silent Mutation | ACA,ACC | T,T 8 | NP_001290370.1 | |
NM_001303442.1 | 1143 | Silent Mutation | ACA,ACC | T,T 8 | NP_001290371.1 | |
NM_001303443.1 | 1143 | Silent Mutation | ACA,ACC | T,T 8 | NP_001290372.1 | |
NM_001303444.1 | 1143 | Silent Mutation | ACA,ACC | T,T 8 | NP_001290373.1 | |
NM_144608.2 | 1143 | Silent Mutation | ACA,ACC | T,T 8 | NP_653209.1 | |
XM_006721687.3 | 1143 | Silent Mutation | ACA,ACC | T,T 31 | XP_006721750.1 | |
XM_011524306.2 | 1143 | Silent Mutation | ACA,ACC | T,T 8 | XP_011522608.1 | |
XM_011524307.2 | 1143 | Silent Mutation | ACA,ACC | T,T 168 | XP_011522609.2 | |
XM_011524308.2 | 1143 | Silent Mutation | ACA,ACC | T,T 169 | XP_011522610.2 | |
XM_017024167.1 | 1143 | Silent Mutation | ACA,ACC | T,T 8 | XP_016879656.1 | |
XM_017024168.1 | 1143 | Silent Mutation | ACA,ACC | T,T 8 | XP_016879657.1 | |
XM_017024169.1 | 1143 | Silent Mutation | ACA,ACC | T,T 8 | XP_016879658.1 | |
XM_017024170.1 | 1143 | Silent Mutation | ACA,ACC | T,T 8 | XP_016879659.1 |
LOC105371795 - uncharacterized LOC105371795 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |