Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCATCTGTTTAGGATCTTTATGCAA[G/C]TCTCTACTGGATTAAAATAAGTGCT
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 605128 | |||||||||||||||||||||||
Literature Links: |
GUCA1C PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
GUCA1C - guanylate cyclase activator 1C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005459.3 | 849 | UTR 3 | NP_005450.3 | |||
XM_005247910.3 | 849 | UTR 3 | XP_005247967.1 | |||
XM_011513334.2 | 849 | UTR 3 | XP_011511636.1 |