Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAGCACTTTGAGGACATGGCCTCC[A/G]ACGAGGACATGTGAGTTGGGGTCTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603265 MIM: 604161 | ||||||||||||||||||||
Literature Links: |
ARID3A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ARID3A - AT-rich interaction domain 3A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005224.2 | 813 | Missense Mutation | AAC,GAC | N,D 120 | NP_005215.1 | |
XM_005259513.4 | 813 | Missense Mutation | AAC,GAC | N,D 120 | XP_005259570.1 | |
XM_005259514.4 | 813 | Missense Mutation | AAC,GAC | N,D 120 | XP_005259571.1 | |
XM_017026445.1 | 813 | Missense Mutation | AAC,GAC | N,D 120 | XP_016881934.1 | |
XM_017026446.1 | 813 | Intron | XP_016881935.1 | |||
XM_017026447.1 | 813 | UTR 5 | XP_016881936.1 |
KISS1R - KISS1 receptor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |