Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATGATTTTGGTGATGGCTGCCTTT[A/G]CATCCCTGTTTCTCAGCGTGTAAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
OR2AT4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
OR2AT4 - olfactory receptor family 2 subfamily AT member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001005285.1 | 2541 | Missense Mutation | GCA,GTA | A,V 302 | NP_001005285.1 | |
XM_017017655.1 | 2541 | Missense Mutation | GCA,GTA | A,V 302 | XP_016873144.1 | |
XM_017017656.1 | 2541 | Missense Mutation | GCA,GTA | A,V 302 | XP_016873145.1 | |
XM_017017657.1 | 2541 | Missense Mutation | GCA,GTA | A,V 302 | XP_016873146.1 | |
XM_017017658.1 | 2541 | Missense Mutation | GCA,GTA | A,V 302 | XP_016873147.1 |