Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAGTGATTATGCTGATTACTTAGA[C/G]AAAGCTGATAAAAACCTGTAGGAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606979 MIM: 602027 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
COG8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
COG8 - component of oligomeric golgi complex 8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NIP7 - NIP7, nucleolar pre-rRNA processing protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TERF2 - telomeric repeat binding factor 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMED6 - transmembrane p24 trafficking protein 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_144676.3 | 412 | Missense Mutation | TCT,TGT | S,C 119 | NP_653277.2 |