Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGGACTACTTGAGACAATGCTGGC[A/G]TCATGATATGCCATACTGGAGCTTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612660 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RFX7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RFX7 - regulatory factor X7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022841.5 | 4340 | Silent Mutation | GAC,GAT | D,D 1037 | NP_073752.5 | |
XM_005254603.3 | 4340 | Silent Mutation | GAC,GAT | D,D 1037 | XP_005254660.2 | |
XM_011521925.2 | 4340 | Silent Mutation | GAC,GAT | D,D 940 | XP_011520227.1 | |
XM_017022506.1 | 4340 | Silent Mutation | GAC,GAT | D,D 1037 | XP_016877995.1 | |
XM_017022507.1 | 4340 | Silent Mutation | GAC,GAT | D,D 940 | XP_016877996.1 | |
XM_017022508.1 | 4340 | Silent Mutation | GAC,GAT | D,D 940 | XP_016877997.1 |