Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCGGCCGACGCCTTCCGCGGCCTG[C/T]GGCGCCTGCGCACGCTCAACCTGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300278 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NYX PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NYX - nyctalopin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022567.2 | 858 | Missense Mutation | CGG,TGG | R,W 254 | NP_072089.1 | |
XM_017029709.1 | 858 | Missense Mutation | CGG,TGG | R,W 254 | XP_016885198.1 |