Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTCCGAAGTCTGGTCTGTGTGGTC[C/T]CTGCTAGCTGAGGCCAGGCCCTGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603730 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SPHK1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SPHK1 - sphingosine kinase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
UBE2O - ubiquitin conjugating enzyme E2 O | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022066.3 | 3613 | Silent Mutation | AGA,AGG | R,R 1213 | NP_071349.3 | |
XM_005257567.1 | 3613 | Silent Mutation | AGA,AGG | R,R 1214 | XP_005257624.1 | |
XM_017024934.1 | 3613 | Silent Mutation | AGA,AGG | R,R 1189 | XP_016880423.1 | |
XM_017024935.1 | 3613 | Silent Mutation | AGA,AGG | R,R 1188 | XP_016880424.1 |