Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAATCGATTTTCAGGTTGTACAGT[C/T]GAGCAGTTTTGGTGGAGAGGGGGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300253 MIM: 300564 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GPR173 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GPR173 - G protein-coupled receptor 173 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TSPYL2 - TSPY like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022117.3 | 582 | Missense Mutation | TCG,TTG | S,L 137 | NP_071400.1 | |
XM_006724592.3 | 582 | Missense Mutation | TCG,TTG | S,L 137 | XP_006724655.1 | |
XM_017029726.1 | 582 | Missense Mutation | TCG,TTG | S,L 137 | XP_016885215.1 | |
XM_017029727.1 | 582 | Missense Mutation | TCG,TTG | S,L 137 | XP_016885216.1 |