Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGCCCAGGATCTCATGGTACATCC[A/G]GAAGTCCTCCTCCTTTCCCCGCAGG
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 608116 | |||||||||||||||||||||||
Literature Links: |
CCDC13 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
CCDC13 - coiled-coil domain containing 13 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_144719.3 | 1550 | Missense Mutation | CGG,TGG | R,W 690 | NP_653320.3 | |
XM_005264898.3 | 1550 | Missense Mutation | CGG,TGG | R,W 649 | XP_005264955.1 | |
XM_005264899.4 | 1550 | Intron | XP_005264956.1 | |||
XM_011533418.2 | 1550 | Missense Mutation | CGG,TGG | R,W 621 | XP_011531720.1 | |
XM_011533419.2 | 1550 | Missense Mutation | CGG,TGG | R,W 470 | XP_011531721.1 | |
XM_011533420.2 | 1550 | Intron | XP_011531722.1 | |||
XM_011533421.2 | 1550 | Intron | XP_011531723.1 | |||
XM_017005781.1 | 1550 | Missense Mutation | CGG,TGG | R,W 470 | XP_016861270.1 |
HHATL - hedgehog acyltransferase-like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |