Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGAAGAGAGAGATCACAATTTTAT[A/G]CATTGTACTGAGAAATAAAAGGCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 217030 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CFI PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CFI - complement factor I | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000204.4 | 1981 | Missense Mutation | GCA,GTA | A,V 583 | NP_000195.2 | |
NM_001318057.1 | 1981 | Missense Mutation | GCA,GTA | A,V 591 | NP_001304986.1 | |
XM_005262976.1 | 1981 | Missense Mutation | GCA,GTA | A,V 576 | XP_005263033.1 | |
XM_006714209.1 | 1981 | Missense Mutation | GCA,GTA | A,V 590 | XP_006714272.1 | |
XM_006714210.3 | 1981 | Intron | XP_006714273.1 | |||
XM_011531920.1 | 1981 | Intron | XP_011530222.1 | |||
XM_017008164.1 | 1981 | Intron | XP_016863653.1 | |||
XM_017008165.1 | 1981 | Intron | XP_016863654.1 | |||
XM_017008166.1 | 1981 | Intron | XP_016863655.1 |